4-Fluoronaphthalene-1-carbonitrile, 97%, Thermo Scientific Chemicals
4-Fluoronaphthalene-1-carbonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoronaphthalene-1-carbonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25335.03
also known as B25335-03
Price (USD)/ Each
50.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
50.30
Add to cart
4-Fluoronaphthalene-1-carbonitrile, 97%, Thermo Scientific Chemicals
Catalog numberB25335.03
Price (USD)/ Each
50.30
-
Add to cart
Chemical Identifiers
CAS13916-99-9
IUPAC Name4-fluoronaphthalene-1-carbonitrile
Molecular FormulaC11H6FN
InChI KeyXQSGYYNFKIQWAC-UHFFFAOYSA-N
SMILESFC1=CC=C(C#N)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Pale yellow to brown
Melting Point (clear melt)86.5-92.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only