Thermo Scientific Chemicals

Ethyl 1H-pyrazole-4-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: B25351.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 1H-pyrazole-4-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: B25351.06
5 g, Each
Quantity
Catalog number: B25351.06
also known as B25351-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37622-90-5
IUPAC Name
ethyl 1H-pyrazole-4-carboxylate
Molecular Formula
C6H8N2O2
InChI Key
KACZQOKEFKFNDB-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CNN=C1
Assay (GC)
≥97.5%
Appearance (Color)
White to pale yellow or pale cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
74.5-80.5?C

Description

Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 1H-pyrazole-4-carboxylate is used in the preparation of isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides. Further, it acts as an intermediate in organic synthesis.

Solubility
Miscible with acetone.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text