2,4-Dimethoxy-1-nitrobenzene, 97%, Thermo Scientific Chemicals
2,4-Dimethoxy-1-nitrobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dimethoxy-1-nitrobenzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B25353.14
also known as B25353-14
Price (USD)/ Each
183.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
183.00
Add to cart
2,4-Dimethoxy-1-nitrobenzene, 97%, Thermo Scientific Chemicals
Catalog numberB25353.14
Price (USD)/ Each
183.00
-
Add to cart
Chemical Identifiers
CAS4920-84-7
IUPAC Name2,4-dimethoxy-1-nitrobenzene
Molecular FormulaC8H9NO4
InChI KeyXXWIYOBCHKCWNT-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(OC)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)71.5-80.5?C
Assay (GC)≥96.0%
Appearance (Color)Pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only