7-Methoxyflavanone, 98%, Thermo Scientific Chemicals
7-Methoxyflavanone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Methoxyflavanone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25358.03
also known as B25358-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS21785-09-1
IUPAC Name7-methoxy-2-phenyl-4H-chromen-4-one
Molecular FormulaC16H12O3
InChI KeyQKNDCRMJDZLFEG-UHFFFAOYSA-N
SMILESCOC1=CC=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Assay (GC)≥97.5%
Appearance (Color)White to pale yellow
Melting Point (clear melt)85.0-91.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only