Thermo Scientific Chemicals

(±)-2-Methyl-2-propanesulfinamide, 97%, Thermo Scientific Chemicals

Catalog number: B25385.03
1 g, Each
Thermo Scientific Chemicals

(±)-2-Methyl-2-propanesulfinamide, 97%, Thermo Scientific Chemicals

Catalog number: B25385.03
1 g, Each
Quantity
Catalog number: B25385.03
also known as B25385-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
146374-27-8
Form
Powder or crystalline powder or lumpy powder
Water Content (Karl Fischer Titration)
<1.0%
Appearance (Color)
White to cream to pale brown
Assay (GC)
≥96.0%

Description

(±)-2-Methyl-2-propanesulfinamide is used in the synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne. It is also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-2-Methyl-2-propanesulfinamide is used in the synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne. It is also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text