Thermo Scientific Chemicals

4-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B25388.06
5 g, Each
Thermo Scientific Chemicals

4-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: B25388.06
5 g, Each
Quantity
Catalog number: B25388.06
also known as B25388-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1711-02-0
IUPAC Name
4-iodobenzoyl chloride
Molecular Formula
C7H4ClIO
InChI Key
NJAKCIUOTIPYED-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(I)C=C1
Melting Point (clear melt)
61.0-68.0?C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.

Solubility
Soluble in toluene and benzene.

Notes
Moisture sensitive. Incompatible with strong bases and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text