Thermo Scientific Chemicals

(Iodomethyl)cyclopentane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B25391.14
25 g, Each
Thermo Scientific Chemicals

(Iodomethyl)cyclopentane, 98+%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B25391.14
25 g, Each
Quantity
Catalog number: B25391.14
also known as B25391-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27935-87-1
IUPAC Name
(iodomethyl)cyclopentane
Molecular Formula
C6H11I
InChI Key
DUMSKQUKLVSSII-UHFFFAOYSA-N
SMILES
ICC1CCCC1
Appearance (Color)
Clear colorless to pale yellow to orange
Assay (GC)
≥98.0%
Form
Liquid
Refractive Index
1.5345-1.5385 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text