1-Boc-isonipecotic acid ethyl ester, 97+%, Thermo Scientific Chemicals
1-Boc-isonipecotic acid ethyl ester, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-isonipecotic acid ethyl ester, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25393.06
also known as B25393-06
Price (USD)/ Each
68.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
68.10
Add to cart
1-Boc-isonipecotic acid ethyl ester, 97+%, Thermo Scientific Chemicals
Catalog numberB25393.06
Price (USD)/ Each
68.10
-
Add to cart
Chemical Identifiers
CAS142851-03-4
IUPAC Name1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate
Molecular FormulaC13H23NO4
InChI KeyMYHJCTUTPIKNAT-UHFFFAOYSA-N
SMILESCCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only