Thermo Scientific Chemicals

Methyl 4-iodo-3-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B25395.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-iodo-3-methylbenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B25395.14
25 g, Each
Quantity
Catalog number: B25395.14
also known as B25395-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5471-81-8
IUPAC Name
methyl 4-iodo-3-methylbenzoate
Molecular Formula
C9H9IO2
InChI Key
HCSGWQGKCVQIRM-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(I)C(C)=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Appearance (Color)
Cream to pale orange
Melting Point (clear melt)
58.0-65.0?C

Description

Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-iodo-3-methylbenzoate is used in the synthesis of novel colchicine-derived nitrate esters as immunosuppressant agents.

Solubility
Slightly soluble in methanol.

Notes
Light sensitive. Store away from oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text