Thermo Scientific Chemicals

Cyclobutylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B25400.06
5 g, Each
Thermo Scientific Chemicals

Cyclobutylamine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B25400.06
5 g, Each
Quantity
Catalog number: B25400.06
also known as B25400-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6291-01-6
IUPAC Name
hydrogen cyclobutanamine chloride
Molecular Formula
C4H10ClN
InChI Key
NFAZOGXQOWEWBM-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1CCC1
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0%
Melting Point (clear melt)
179.0-188.0?C
Appearance (Color)
White

Description

Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry conditions, in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text