Thermo Scientific Chemicals

Papaverine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: B25412.06
5 g, Each
Thermo Scientific Chemicals

Papaverine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: B25412.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B25412.06
also known as B25412-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61-25-6
IUPAC Name
hydrogen 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline chloride
Molecular Formula
C20H22ClNO4
InChI Key
UOTMYNBWXDUBNX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].COC1=CC=C(CC2=C3C=C(OC)C(OC)=CC3=CC=N2)C=C1OC
Form
powder
Appearance (Color)
White to yellow
Assay (Titration ex Chloride)
> 98.5%

Description

Papaverine hydrochloride is used as a smooth muscle relaxant, with noted treatment for impotence. It can also act as a cerebral vasodilator through the possible inhibition of PDE (phosphodiesterases) and/or calcium channel actions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (50 mg/ml), DMSO, methanol, 100% ethanol and chloroform.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text