Thermo Scientific Chemicals

(+/-)-Boc-alpha-phosphonoglycine trimethyl ester, 95%, Thermo Scientific Chemicals

Catalog number: B25421.14
25 g, Each
Thermo Scientific Chemicals

(+/-)-Boc-alpha-phosphonoglycine trimethyl ester, 95%, Thermo Scientific Chemicals

Catalog number: B25421.14
25 g, Each
Quantity
Catalog number: B25421.14
also known as B25421-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89524-98-1
IUPAC Name
methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular Formula
C10H20NO7P
InChI Key
LJHAPRKTPAREGO-ZETCQYMHSA-N
SMILES
COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC
Appearance (Color)
White to cream or pale yellow
Form
Powder
Assay (GC)
≥94.0%
Elemental Analysis
C: 40.11 - 40.71% (Theory 40.41%)
Elemental Analysis
H: 6.48 - 7.08% (Theory 6.78%)

Description

It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text