2-Formylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
2-Formylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Formylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25434.03
also known as B25434-03
Price (USD)/ Each
33.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
33.20
Add to cart
2-Formylbenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberB25434.03
Price (USD)/ Each
33.20
-
Add to cart
Chemical Identifiers
CAS40138-16-7
IUPAC Name(2-formylphenyl)boronic acid
Molecular FormulaC7H7BO3
InChI KeyDGUWACLYDSWXRZ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale pink
Assay (HPLC)≥96.0%
Melting Point (clear melt)119.0-129.0?C
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
View more
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Alois Furstner; Victor Mamane. Concise total synthesis of the aporphine alkaloid 7,7'-bisdehydro-O-methylisopiline by an InCl3 mediated cycloisomerization reaction. Chem. Commun. 2003, (17), 2112-2113. 
  2. Christopher J.Ward; Prakash Patel; Tony D.James. Boronic acid appended azo dyes-color sensors for saccharides. J. Chem. Soc., Perkin Trans. 1. 2002, (4), 462-470.
  3. Used in combination with (R)- or (S)-1,1'-bi-2-naphthol in a protocol for determining the enantiomeric excess of primary amines by NMR: Org. Lett., 8, 609 (2006).