Thermo Scientific Chemicals

1-Methylbenzimidazole, 97%, Thermo Scientific Chemicals

Catalog number: B25472.14
25 g, Each
Thermo Scientific Chemicals

1-Methylbenzimidazole, 97%, Thermo Scientific Chemicals

Catalog number: B25472.14
25 g, Each
Quantity
Catalog number: B25472.14
also known as B25472-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1632-83-3
IUPAC Name
1-methyl-1H-1,3-benzodiazole
Molecular Formula
C8H8N2
InChI Key
FGYADSCZTQOAFK-UHFFFAOYSA-N
SMILES
CN1C=NC2=CC=CC=C12
Appearance (Color)
White to cream to pale brown or pink
Assay (GC)
≥96.0%
Melting Point (clear melt)
55.0-63.0°C
Form
Crystals or powder or crystalline powder

Description

1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text