3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Seongyong Kim; Kohei Imura; Minwoo Lee; Tetsuya Narushima; Hiromi Okamoto; Dae Hong Jeong. Strong optical coupling between mutually orthogonal plasmon oscillations in a silver nanosphere-nanowire joined system. Physical Chemistry Chemical Physics. 2013, 15,(12), 4146-4153.
- Arun Syamal. Syntheses of new oxovanadium(IV) complexes with some new Schiff bases derived from salicylaldehyde or substituted salicylaldehyde and 3-aminothiophenol. Transition Metal Chemistry. 1978, 3,(1), 297-299.