Thermo Scientific Chemicals

2-Bromoindene, 98%, Thermo Scientific Chemicals

Catalog number: B25474.14
25 g, Each
Thermo Scientific Chemicals

2-Bromoindene, 98%, Thermo Scientific Chemicals

Catalog number: B25474.14
25 g, Each
Quantity
Catalog number: B25474.14
also known as B25474-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10485-09-3
IUPAC Name
2-bromo-1H-indene
Molecular Formula
C9H7Br
InChI Key
CCUYEVNCRQDQRF-UHFFFAOYSA-N
SMILES
BrC1=CC2=CC=CC=C2C1
Appearance (Color)
Pale yellow to brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
34.0-40.0?C
Form
Powder or fused solid

Description

2-Bromoindene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoindene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text