Thermo Scientific Chemicals

2-Bromo-1,3-dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: B25485.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-1,3-dimethoxybenzene, 98%, Thermo Scientific Chemicals

Catalog number: B25485.14
25 g, Each
Quantity
Catalog number: B25485.14
also known as B25485-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16932-45-9
IUPAC Name
2-bromo-1,3-dimethoxybenzene
Molecular Formula
C8H9BrO2
InChI Key
VHVYSMMZHORFKU-UHFFFAOYSA-N
SMILES
COC1=CC=CC(OC)=C1Br
Appearance (Color)
Pale cream to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
90.5-96.5?C

Description

2-Bromo-1,3-dimethoxybenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-1,3-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text