4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-(4-Methoxyphenyl)-2,2′:6′,2′′-terpyridine is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Arockiam Anthonysamy; Sengottuvelan Balasubramanian; Vellasamy Shanmugaiah; Narayanasamy Mathivanan. Synthesis, characterization and electrochemistry of 4'-functionalized 2,2':6',2″-terpyridine ruthenium(II) complexes and their biological activity. Dalton Trans. 2008, (16), 2136-2143.
- Yu-Zhe Chen; Deng-Hui Wang; Bin Chen; Jian-Ji Zhong; Chen-Ho Tung; Li-Zhu Wu. Visible light-induced synthesis of 3,4-diarylthiophenes from 3,4-diaryl-2,5-dihydrothiophenes. J. Org. Chem. 2012, 77,(16), 6773-6777.