5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals
5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25496.03
also known as B25496-03
Price (USD)/ Each
40.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
40.70
Add to cart
5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals
Catalog numberB25496.03
Price (USD)/ Each
40.70
-
Add to cart
Chemical Identifiers
CAS499-49-0
IUPAC Name5-methylbenzene-1,3-dicarboxylic acid
Molecular FormulaC9H8O4
InChI KeyPMZBHPUNQNKBOA-UHFFFAOYSA-N
SMILESCC1=CC(=CC(=C1)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)295.0-302.0?C
FormPowder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only