2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B25497.14
also known as B25497-14
Price (USD)/ Each
295.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
295.00
Add to cart
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
Catalog numberB25497.14
Price (USD)/ Each
295.00
-
Add to cart
Chemical Identifiers
CAS98446-49-2
IUPAC Name2,4-dichloro-5-methoxyaniline
Molecular FormulaC7H7Cl2NO
InChI KeyAJROJTARXSATEB-UHFFFAOYSA-N
SMILESCOC1=CC(N)=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormPowder
Appearance (Color)Pale brown to brown
Melting Point (clear melt)47.0-53.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only