4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Philippe Cotellea & Jean Pierre Catteau. Nitrodecarboxylation and Nitrodeformylation of Some Electron-Rich Benzoic Acids and Benzaldehydes. Synthetic Communications. 1996, 26 (22), 4105-4112.
Bruce A. Hathaway; Brian E. Taylor & Jeremy S. Wittenborn. Selective Preparation of 4-(Bromo or Iodo)-2,5-dimethoxybenzonitrile and 2-(Bromo or Iodo)-3,6-dimethoxybenzonitrile from 2,5-Dimethoxybenzaldehyde. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1998, 28 (24), 4629-4637.