Thermo Scientific Chemicals

Phloroglucinol, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: B25502.18
50 g, Each
Thermo Scientific Chemicals

Phloroglucinol, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: B25502.18
50 g, Each
Quantity
Catalog number: B25502.18
also known as B25502-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-73-6
IUPAC Name
benzene-1,3,5-triol
Molecular Formula
C6H6O3
InChI Key
QCDYQQDYXPDABM-UHFFFAOYSA-N
SMILES
OC1=CC(O)=CC(O)=C1
Form
Powder
Melting Point (clear melt)
213-223?C
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥97.5%
Water Content (Karl Fischer Titration)
≤1.5%

Description

Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for pentoses, pentosans and aldehydes. Used as a bone decalcifier in microscopy specimens.Phloroglucinol is mainly used as a coupling agent in printing. It is an active component of Tollen’s reagent and Gunzburg reagent used to test pentoses and hydrochloric acid in gastric juice respectively. In analytical chemistry, it is used to study condensed tannins by means of depolymerization. It is also involved in the synthesis of 2,4,6-triamino-1,3,5- trinitrobenzene, trinitrophloroglucinol and pharmaceuticals like flopropione .

Solubility
Soluble in diethyl ether, ethanol and pyridine.

Notes
Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text