Thermo Scientific Chemicals

Cinnamonitrile, 97%, predominantly trans, Thermo Scientific Chemicals

Catalog number: B25503.14
25 g, Each
Thermo Scientific Chemicals

Cinnamonitrile, 97%, predominantly trans, Thermo Scientific Chemicals

Catalog number: B25503.14
25 g, Each
Quantity
Catalog number: B25503.14
also known as B25503-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1885-38-7
IUPAC Name
(2E)-3-phenylprop-2-enenitrile
Molecular Formula
C9H7N
InChI Key
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
SMILES
N#C\C=C\C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.5990-1.6040 @ 20°C
Form
Liquid
Impurity content
<10% cis-Cinnamonitrile by GC
Assay (GC)
≥96.0% (sum of cis/trans)

Description

Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text