Thermo Scientific Chemicals

Methyl 4-iodo-2-methoxybenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: B25529.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-iodo-2-methoxybenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: B25529.14
25 g, Each
Quantity
Catalog number: B25529.14
also known as B25529-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
148490-97-5
IUPAC Name
methyl 4-iodo-2-methoxybenzoate
Molecular Formula
C9H9IO3
InChI Key
MCNOTXROWOGSGU-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(OC)C=C(I)C=C1
Appearance (Color)
White to cream
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
41.0-48.0?C

Description

Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light, oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text