2-Bromo-6-methoxyphenol, 98+%, Thermo Scientific Chemicals
2-Bromo-6-methoxyphenol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-6-methoxyphenol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25535.03
also known as B25535-03
Price (USD)/ Each
62.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
62.20
Add to cart
2-Bromo-6-methoxyphenol, 98+%, Thermo Scientific Chemicals
Catalog numberB25535.03
Price (USD)/ Each
62.20
-
Add to cart
Chemical Identifiers
CAS28165-49-3
IUPAC Name2-bromo-6-methoxyphenol
Molecular FormulaC7H7BrO2
InChI KeyWEUFQISIJPSTBM-UHFFFAOYSA-N
SMILESCOC1=C(O)C(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)61.0-67.0?C
Appearance (Color)White
2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Ling-Wei Hsin; Li-Te Chang; Chien-Wei Chen; Chia-Huei Hsu; Hung-Wei Chen. Stereoselective synthesis of morphine fragments trans- and cis-octahydro-1H-benzo[4,5]furo[3,2-e]isoquinolines. Tetrahedron. 2005, 61 (2), 513-520.
  2. Ji-Qiang Chen; Jian-Hua Xie; Deng-Hui Bao; Sheng Liu and Qi-Lin Zhou. Total Synthesis of (-)-Galanthamine and (-)-Lycoramine via Catalytic Asymmetric Hydrogenation and Intramolecular Reductive Heck Cyclization. Acta Cryst. 2012, 14 (11), 2714-2717.