1-Bromo-2-chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-2-chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number B25538.06
also known as B25538-06
Price (USD)/ Each
89.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
89.50
Add to cart
1-Bromo-2-chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals
Catalog numberB25538.06
Price (USD)/ Each
89.50
-
Add to cart
Chemical Identifiers
CAS110407-59-5
IUPAC Name1-bromo-2-chloro-4-fluorobenzene
Molecular FormulaC6H3BrClF
InChI KeyLEFQPBAWVJEIJS-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5500-1.5540 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only