Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-iodo-3-nitrobenzoate is used as pharmaceutical intermediate.
Solubility
Soluble in DCM 10mg/0.5mL. Insoluble in water.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
RUO – Research Use Only
F. H. Allen and D. Rogers. A reference list of organic structures whose absolute configurations have been determined by X-ray fluorescence. Chem. Commun. (London). 1966, (22), 838-841.
Koichi Nakazawa. Syntheses of Ring-substituted Flavonoids and Allied Compounds. XI. Synthesis of Hinokiflavone. Chemical and Pharmaceutical Bulletin. 1968, 16 (12), 2503-2511.