Thermo Scientific Chemicals

2-(Chloromethyl)benzimidazole, 95%, Thermo Scientific Chemicals

Catalog number: B25548.14
25 g, Each
Thermo Scientific Chemicals

2-(Chloromethyl)benzimidazole, 95%, Thermo Scientific Chemicals

Catalog number: B25548.14
25 g, Each
Quantity
Catalog number: B25548.14
also known as B25548-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4857-04-9
IUPAC Name
2-(chloromethyl)-1H-1,3-benzodiazole
Molecular Formula
C8H7ClN2
InChI Key
SPMLMLQATWNZEE-UHFFFAOYSA-N
SMILES
ClCC1=NC2=CC=CC=C2N1
Appearance (Color)
White to cream to yellow brown
Assay (Non-aqueous acid-base Titration)
≥94% to ≤106.0%
Assay (HPLC)
≥94.0%
Identification (FTIR)
Conforms
Form
Powder

Description

2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text