Thermo Scientific Chemicals

8-Bromooctanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: B25575.09
10 g, Each
Thermo Scientific Chemicals

8-Bromooctanoic acid, 95%, Thermo Scientific Chemicals

Catalog number: B25575.09
10 g, Each
Quantity
Catalog number: B25575.09
also known as B25575-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17696-11-6
IUPAC Name
8-bromooctanoate
Molecular Formula
C8H14BrO2
InChI Key
BKJFDZSBZWHRNH-UHFFFAOYSA-M
SMILES
[O-]C(=O)CCCCCCCBr
Appearance (Color)
White to pale yellow to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
33.0-42.0?C

Description

8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4'-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4′-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text