8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4'-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Bromooctanoic acid is used in the synthesis of 8-(N-Methyl-4,4′-bipyridinyl)- octanoic acid. 8-Mercaptooctanoic acid was prepared from 8-bromooctanoic acid.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents, bases.
RUO – Research Use Only
Amihood Doron; Eugenii Katz; Itamar Willner. Organization of Au Colloids as Monolayer Films onto ITO Glass Surfaces: Application of the Metal Colloid Films as Base Interfaces To Construct Redox-Active Monolayers. Langmuir. 1995, 11 (4), 1313-1317.
Mark A. Hayden; Irene Y. Huang; Gina Iliopoulos; Marilu Orozco; Gary W. Ashley. Biosynthesis of lipoic acid: Characterization of the lipoic acid auxotrophs Escherichia coli W1485-lip2 and JRG33-lip9. Biochemistry. 1993, 32 (14), 3778-3782.