4-Cyanobenzamide, 97%, Thermo Scientific Chemicals
4-Cyanobenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanobenzamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B25600.06
also known as B25600-06
Price (USD)/ Each
210.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
248.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
210.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
248.00 
Save 37.35 (15%)
Add to cart
4-Cyanobenzamide, 97%, Thermo Scientific Chemicals
Catalog numberB25600.06
Price (USD)/ Each
210.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
248.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
CAS3034-34-2
IUPAC Name4-cyanobenzamide
Molecular FormulaC8H6N2O
InChI KeyFUKWTMJZHKZKFA-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥96.0%
Appearance (Color)White to pale cream
Melting Point (clear melt)223.0-229.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only