Thermo Scientific Chemicals

3-Bromo-4-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: B25610.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-4-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog number: B25610.06
5 g, Each
Quantity
Catalog number: B25610.06
also known as B25610-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
656-64-4
IUPAC Name
3-bromo-4-fluoroaniline
Molecular Formula
C6H5BrFN
InChI Key
KOWPUNQBGWIERF-UHFFFAOYSA-N
SMILES
NC1=CC(Br)=C(F)C=C1
Appearance (Color)
Pale cream to cream to yellow to gray
Form
Crystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
27.0-34.0?C

Description

3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text