5-Fluoro-2-methylanisole, 98%, Thermo Scientific Chemicals
5-Fluoro-2-methylanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluoro-2-methylanisole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25629.03
also known as B25629-03
Price (USD)/ Each
28.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
28.00
Add to cart
5-Fluoro-2-methylanisole, 98%, Thermo Scientific Chemicals
Catalog numberB25629.03
Price (USD)/ Each
28.00
-
Add to cart
Chemical Identifiers
CAS95729-22-9
IUPAC Name4-fluoro-2-methoxy-1-methylbenzene
Molecular FormulaC8H9FO
InChI KeyCXTIUQIZWCABPV-UHFFFAOYSA-N
SMILESCOC1=CC(F)=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4920-1.4960 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only