Phenanthrene-9-carboxaldehyde, 97%, Thermo Scientific Chemicals
Phenanthrene-9-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenanthrene-9-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25648.03
also known as B25648-03
Price (USD)/ Each
56.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
56.30
Add to cart
Phenanthrene-9-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberB25648.03
Price (USD)/ Each
56.30
-
Add to cart
Chemical Identifiers
CAS4707-71-5
IUPAC Namephenanthrene-9-carbaldehyde
Molecular FormulaC15H10O
InChI KeyQECIGCMPORCORE-UHFFFAOYSA-N
SMILESO=CC1=CC2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale yellow
FormPowder
Assay (GC)≥96.0%
Free acid (titration)≤2.25%
Identification (FTIR)Conforms
View more
Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

Solubility
Soluble in acetone, dichloromethane, methanol.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
RUO – Research Use Only

General References:

  1. Thomas J. Katz; Timothy M. Sivavec. Metal-catalyzed rearrangement of alkene-alkynes and the stereochemistry of metallacyclobutene ring opening. J. Am. Chem. Soc. 1985, 107 (3), 737-738.
  2. K.A Fähnrich; M Pravda; G.G Guilbault. Disposable amperometric immunosensor for the detection of polycyclic aromatic hydrocarbons (PAHs) using screen-printed electrodes. Biosensors and Bioelectronics. 2003, 18 (1), 73-82.