Thermo Scientific Chemicals

Phenanthrene-9-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B25648.06
5 g, Each
Thermo Scientific Chemicals

Phenanthrene-9-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: B25648.06
5 g, Each
Quantity
Catalog number: B25648.06
also known as B25648-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4707-71-5
IUPAC Name
phenanthrene-9-carbaldehyde
Molecular Formula
C15H10O
InChI Key
QECIGCMPORCORE-UHFFFAOYSA-N
SMILES
O=CC1=CC2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
Pale cream to pale yellow
Form
Powder
Assay (GC)
≥96.0%
Free acid (titration)
≤2.25%
Identification (FTIR)
Conforms

Description

Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

Solubility
Soluble in acetone, dichloromethane, methanol.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text