(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.
Solubility
Soluble in water, 4.526 mg/L @ 25°C.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- M. Venkat Ram Reddy; Ahmet J. Yucel and P. Veeraraghavan Ramachandran. An Efficient Asymmetric Synthesis of Tarchonanthuslactone. J. Org. Chem. 2001, 66 (7), 2512-2514.
- Christopher W. G. Au; Robert J. Nashb and Stephen G. Pyne. Synthesis of hyacinthacine B3 and purported hyacinthacine B7. Chem. Commun. 2010, 46 (5), 713-715.