Thermo Scientific Chemicals

(R)-(-)-4-Penten-2-ol, 95%, Thermo Scientific Chemicals

Catalog number: B25659.06
5 g, Each
Thermo Scientific Chemicals

(R)-(-)-4-Penten-2-ol, 95%, Thermo Scientific Chemicals

Catalog number: B25659.06
5 g, Each
Quantity
Catalog number: B25659.06
also known as B25659-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64584-92-5
IUPAC Name
pent-4-en-2-ol
Molecular Formula
C5H10O
InChI Key
ZHZCYWWNFQUZOR-UHFFFAOYNA-N
SMILES
CC(O)CC=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥94.0%
Optical Rotation
-4.7 ± 0.1? (neat)
Refractive Index
1.4205-1.4265 @ 20?C

Description

(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-4-Penten-2-ol is used as a pharmaceutical intermediate.

Solubility
Soluble in water, 4.526 mg/L @ 25°C.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text