4'-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Aminobenzanilide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field. It has anticonvulsant property.
Solubility
Soluble in DMSO and methanol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- C. Randall Clark; Ricky T. Sansom; Ching Ming Lin; George N. Norris. Anticonvulsant activity of some 4-aminobenzanilides. J. Med. Chem. 1985, 28 (9), 1259-1262.
- Paul A. Wender; Sofia M. Touami; Carole Alayrac and Ulrich C. Philipp. Triazole Photonucleases: A New Family of Light Activatable DNA Cleaving Agents. J. Am. Chem. Soc. 1996, 118 (27), 6522-6523.