1,5-Pentanedithiol, 96%, Thermo Scientific Chemicals
1,5-Pentanedithiol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,5-Pentanedithiol, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B25665.14
also known as B25665-14
Price (USD)/ Each
192.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
226.00 
Save 33.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
192.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
226.00 
Save 33.35 (15%)
Add to cart
1,5-Pentanedithiol, 96%, Thermo Scientific Chemicals
Catalog numberB25665.14
Price (USD)/ Each
192.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
226.00 
Save 33.35 (15%)
-
Add to cart
Chemical Identifiers
CAS928-98-3
IUPAC Namepentane-1,5-dithiol
Molecular FormulaC5H12S2
InChI KeyKMTUBAIXCBHPIZ-UHFFFAOYSA-N
SMILESSCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5180-1.5240 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only