Thermo Scientific Chemicals

4-Bromobenzenediazonium tetrafluoroborate, 96%, Thermo Scientific Chemicals

Catalog number: B25670.06
5 g, Each
Thermo Scientific Chemicals

4-Bromobenzenediazonium tetrafluoroborate, 96%, Thermo Scientific Chemicals

Catalog number: B25670.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B25670.06
also known as B25670-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
673-40-5
IUPAC Name
4-bromobenzene-1-diazonium; tetrafluoroboranuide
Molecular Formula
C6H4BBrF4N2
InChI Key
FXTCQUCYDJZGGU-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N
Appearance (Color)
Pale cream to pale brown or pale pink
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%

Description

4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text