Thermo Scientific Chemicals

2,6-Diacetylpyridine, 99%, Thermo Scientific Chemicals

Catalog number: B25678.14
25 g, Each
Thermo Scientific Chemicals

2,6-Diacetylpyridine, 99%, Thermo Scientific Chemicals

Catalog number: B25678.14
25 g, Each
Quantity
Catalog number: B25678.14
also known as B25678-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1129-30-2
IUPAC Name
1-(6-acetylpyridin-2-yl)ethan-1-one
Molecular Formula
C9H9NO2
InChI Key
BEZVGIHGZPLGBL-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC(=N1)C(C)=O
Appearance (Color)
White to cream
Assay (GC)
≥98.5%
Melting Point (clear melt)
77.0-84.0°C
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms

Description

2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.

Solubility
Soluble in water, chloroform, and DMSO.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text