Methyl 4-fluoro-3-hydroxybenzoate, 98+%, Thermo Scientific Chemicals
Methyl 4-fluoro-3-hydroxybenzoate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-fluoro-3-hydroxybenzoate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25683.06
also known as B25683-06
Price (USD)/ Each
75.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
75.30
Add to cart
Methyl 4-fluoro-3-hydroxybenzoate, 98+%, Thermo Scientific Chemicals
Catalog numberB25683.06
Price (USD)/ Each
75.30
-
Add to cart
Chemical Identifiers
CAS214822-96-5
IUPAC Namemethyl 4-fluoro-3-hydroxybenzoate
Molecular FormulaC8H7FO3
InChI KeyCUGWNEOTLGLGDG-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(O)=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥98.0%
Melting Point (clear melt)89.0-95.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only