Thermo Scientific Chemicals

Methyl 4-fluoro-3-hydroxybenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: B25683.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-fluoro-3-hydroxybenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: B25683.14
25 g, Each
Quantity
Catalog number: B25683.14
also known as B25683-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
214822-96-5
IUPAC Name
methyl 4-fluoro-3-hydroxybenzoate
Molecular Formula
C8H7FO3
InChI Key
CUGWNEOTLGLGDG-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(O)=C(F)C=C1
Appearance (Color)
White to pale cream
Assay (GC)
≥98.0%
Melting Point (clear melt)
89.0-95.0?C
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text