2',6'-Dimethylacetanilide, 97%, Thermo Scientific Chemicals
2',6'-Dimethylacetanilide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2',6'-Dimethylacetanilide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B25687.14
also known as B25687-14
Price (USD)/ Each
39.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
39.30
Add to cart
2',6'-Dimethylacetanilide, 97%, Thermo Scientific Chemicals
Catalog numberB25687.14
Price (USD)/ Each
39.30
-
Add to cart
Chemical Identifiers
CAS2198-53-0
IUPAC NameN-(2,6-dimethylphenyl)acetamide
Molecular FormulaC10H13NO
InChI KeyNRPTXWYBRKRZES-UHFFFAOYSA-N
SMILESCC(=O)NC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or lumps
Assay (GC)≥96.0%
Melting Point (clear melt)178.0-184.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only