Thermo Scientific Chemicals

1-Bromo-2-isopropoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: B25688.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-2-isopropoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: B25688.14
25 g, Each
Quantity
Catalog number: B25688.14
also known as B25688-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
701-07-5
IUPAC Name
1-bromo-2-(propan-2-yloxy)benzene
Molecular Formula
C9H11BrO
InChI Key
MMORVPBHAHXAHH-UHFFFAOYSA-N
SMILES
CC(C)OC1=CC=CC=C1Br
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5355-1.5405 @ 20?C

Description

1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-isopropoxybenzene is used as an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text