Thermo Scientific Chemicals

5-Bromo-2-methoxyaniline, 97%, Thermo Scientific Chemicals

Catalog number: B25693.06
5 g, Each
Thermo Scientific Chemicals

5-Bromo-2-methoxyaniline, 97%, Thermo Scientific Chemicals

Catalog number: B25693.06
5 g, Each
Quantity
Catalog number: B25693.06
also known as B25693-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6358-77-6
IUPAC Name
5-bromo-2-methoxyaniline
Molecular Formula
C7H8BrNO
InChI Key
OPGNSNDTPPIYPG-UHFFFAOYSA-N
SMILES
COC1=C(N)C=C(Br)C=C1
Appearance (Color)
White to cream to pale brown or pale grey
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
92.5-101.5?C

Description

5-Bromo-2-methoxyaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-methoxyaniline is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Preserve in well- closed, light- resistant & tight container. Store in cool & dry place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text