3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chlorpropionyl chloride can be used in the production process of pharma ingredients.
Solubility
Reacts violently with water.
Notes
Moisture sensitive. Store away from oxidizing agents, water/ mois
RUO – Research Use Only
General References:
- Henry M. Miziorko; Christine E. Behnke. Active-site-directed inhibition of 3-hydroxy-3-methylglutaryl coenzyme A synthase by 3-chloropropionyl coenzyme A. Biochemistry. 1974, 24 (13), 3174-3179.
- Hassan M. Badawi; Wolfgang Förner. The potential function for the internal rotation and the derived potential energy distribution of the normal modes for 3-chloropropionyl chloride based on ab initio calculations. Journal of Molecular Structure: THEOCHEM. 1999, 488 (1-3), 69-85.