Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
RUO – Research Use Only
Carolin Rether and Carsten Schmuck. Carboxylate Binding by Indole-Based Guanidinium Receptors: Acylguanidinium Cations are Better than Aromatic Guanidinium Cations. European Journal of Organic Chemistry. 2011, 2001 (8), 1459-1466.
Dr. Jyotirmayee Dash; Rabindra Nath Das; Nagaratna Hegde; Dr. G. Dan Panto; Pravin S. Shirude; Prof.Dr. Shankar Balasubramanian. Synthesis of Bis-indole Carboxamides as G-Quadruplex Stabilizing and Inducing Ligands. Chemistry an European Journal. 2012, 18 (2), 554-564.