Thermo Scientific Chemicals

Methyl 3-amino-4-iodobenzoate, 97%, Thermo Scientific Chemicals

Catalog number: B25699.06
5 g, Each
Thermo Scientific Chemicals

Methyl 3-amino-4-iodobenzoate, 97%, Thermo Scientific Chemicals

Catalog number: B25699.06
5 g, Each
Quantity
Catalog number: B25699.06
also known as B25699-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
412947-54-7
IUPAC Name
methyl 3-amino-4-iodobenzoate
Molecular Formula
C8H8INO2
InChI Key
WJEBNIVVLJEIKE-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(N)=C(I)C=C1
Assay (GC)
≥96.0%
Melting Point (clear melt)
130.0-140.0?C
Form
Powder
Appearance (Color)
Yellow

Description

Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-amino-4-iodobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text