Thermo Scientific Chemicals

1-Aminopyridinium iodide, 97%, Thermo Scientific Chemicals

Catalog number: B25703.14
25 g, Each
Thermo Scientific Chemicals

1-Aminopyridinium iodide, 97%, Thermo Scientific Chemicals

Catalog number: B25703.14
25 g, Each
Quantity
Catalog number: B25703.14
also known as B25703-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6295-87-0
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Titration ex Iodide)
≥96.0 to ≤104.0%

Description

1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.

Solubility
Soluble in water (almost transparency).

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text